Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009208.3 | NP_033234.2 | 13 - 14 | 2183 | 156 | |
GenBank mRNA | AK147218.1 | - | 2520 | 156 | ||
BC066086.1 | - | 14 - 15 | 2057 | 156 | ||
BC080271.1 | - | 13 - 14 | 2178 | 6831942 | 156 | |
M28383.1 | - | 13 - 14 | 2153 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |