Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK002399.1 | - | 7 - 8 | 604 | 68 | |
AK011171.1 | - | 4 - 5 | 312 | 68 | ||
AK171752.1 | - | 7 - 8 | 581 | 68 | ||
BC022715.1 | - | 7 - 8 | 569 | 68 | ||
BC048821.1 | - | 7 - 8 | 591 | 3966642 | 68 | |
BE853767.1 | - | 1 - 2 | 76 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |