Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133708.3 | NP_598469.1 | 9 - 10 | 941 | 71 | |
GenBank mRNA | AK144311.1 | - | 9 - 10 | 1206 | 71 | |
AK150473.1 | - | 10 - 11 | 1077 | 71 | ||
BC008116.1 | - | 9 - 10 | 900 | 3498021 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |