Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029010.1 | NP_083286.1 | 15 - 16 | 1714 | 95 | |
GenBank mRNA | AK014667.1 | - | 13 - 14 | 1737 | 95 | |
AK029925.1 | - | 15 - 16 | 1714 | 95 | ||
BC021773.1 | - | 14 - 15 | 1467 | 4222994 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |