Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177222.4 | NP_796196.3 | 9 - 10 | 1188 | 110 | |
GenBank mRNA | AK132912.1 | - | 9 - 10 | 1412 | 110 | |
AY423542.1 | - | 9 - 10 | 1119 | 110 | ||
AY485607.1 | - | 9 - 10 | 1188 | 110 | ||
AY485608.1 | - | 9 - 10 | 1188 | 110 | ||
BC072568.1 | - | 7 - 8 | 1030 | 110 | ||
BC120743.1 | - | 9 - 10 | 1092 | 40129666 | 110 | |
BC125423.1 | - | 9 - 10 | 1092 | 40129938 | 110 | |
BC144801.1 | - | 9 - 10 | 1092 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |