Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bspry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138653.1 | NP_619594.1 | 1 - 2 | 367 | 100 | |
GenBank mRNA | AJ276690.1 | - | 1 - 2 | 367 | 100 | |
AK035588.1 | - | 1 - 2 | 373 | 100 | ||
AK160930.1 | - | 1 - 2 | 373 | 100 | ||
BC139079.1 | - | 1 - 2 | 383 | 8862101 | 100 | |
BC139080.1 | - | 1 - 2 | 383 | 8862102 | 100 | |
BC145535.1 | - | 1 - 2 | 383 | 9054098 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |