Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025811.3 | NP_080087.1 | 1 - 2 | 596 | 66 | |
GenBank mRNA | AK002650.1 | - | 1 - 2 | 181 | 66 | |
AK004564.1 | - | 1 - 2 | 243 | 66 | ||
AK033384.1 | - | 1 - 2 | 597 | 66 | ||
BC095956.1 | - | 1 - 2 | 540 | 5330002 | 66 | |
BC138190.1 | - | 1 - 2 | 433 | 8861209 | 66 | |
BC138191.1 | - | 1 - 2 | 433 | 8861210 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |