Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025811.3 | NP_080087.1 | 5 - 6 | 1460 | 99 | |
GenBank mRNA | AK002650.1 | - | 5 - 6 | 1045 | 99 | |
AK004564.1 | - | 5 - 6 | 1107 | 99 | ||
AK033384.1 | - | 5 - 6 | 1461 | 99 | ||
BC041104.1 | - | 3 - 4 | 429 | 99 | ||
BC050211.1 | - | 3 - 4 | 316 | 99 | ||
BC095956.1 | - | 5 - 6 | 1404 | 5330002 | 99 | |
BC138190.1 | - | 5 - 6 | 1297 | 8861209 | 99 | |
BC138191.1 | - | 5 - 6 | 1297 | 8861210 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |