Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013913.3 | NP_038941.1 | 2 - 3 | 653 | 112 | |
GenBank mRNA | AF162224.1 | - | 2 - 3 | 621 | 112 | |
AK149450.1 | - | 2 - 3 | 654 | 112 | ||
AK149555.1 | - | 2 - 3 | 654 | 112 | ||
BC019491.1 | - | 2 - 3 | 641 | 4211688 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |