Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145550.3 | NP_663525.1 | 4 - 5 | 425 | 88 | |
GenBank mRNA | AK081205.1 | - | 4 - 5 | 412 | 88 | |
AK157536.1 | - | 4 - 5 | 412 | 88 | ||
BC009080.1 | - | 4 - 5 | 357 | 2655774 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |