Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctex1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163767.1 | NP_001157239.1 | 2 - 3 | 324 | 77 | |
NM_026100.3 | NP_080376.1 | 2 - 3 | 356 | 77 | ||
GenBank mRNA | AK006804.1 | - | 2 - 3 | 325 | 77 | |
AK015315.1 | - | 2 - 3 | 359 | 77 | ||
BC048635.1 | - | 2 - 3 | 296 | 77 | ||
BC099549.1 | - | 2 - 3 | 330 | 30919464 | 77 | |
BC132398.1 | - | 2 - 3 | 274 | 40130675 | 77 | |
BC132400.1 | - | 2 - 3 | 274 | 40130677 | 77 | |
BC145187.1 | - | 2 - 3 | 274 | 9053738 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |