Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
BC025810.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
AK169290.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
AY535001.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
AK160070.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
BC017637.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
BC025810.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
AK169290.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
AY535001.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
AK160070.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208580 |
BC017637.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
12027 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
272787 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
456953 |
AK017808.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
456953 |
AK079384.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
456953 |
BC018493.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
456953 |
NM_027495.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AF006482.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AK002618.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AK031581.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AK036641.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AK045828.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AK080265.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AK081435.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AK088455.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AK160950.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
AK163688.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
BC015247.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
NM_001026214.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923311 |
NM_007647.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
663885 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
289481 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
42498 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
64030 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P137256 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P164703 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P165221 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P127311 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
BC025810.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
AK169290.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
AY535001.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
AK160070.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
BC017637.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
BC025810.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
AK169290.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
AY535001.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
AK160070.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208580 |
BC017637.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006711 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006736 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006727 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006722 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006714 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006765 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006732 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006763 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006735 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006712 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006733 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006710 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006718 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006729 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006713 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006716 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006723 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006748 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006746 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20006715 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09518448 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09518458 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P137256 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09518466 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09518447 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09518471 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03807730 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P164703 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P165221 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P405265 |
BC025810.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P405265 |
BY283382.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P405265 |
AK169290.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P405265 |
AY535001.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P405265 |
AK160070.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
BC025810.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
AK169290.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
AY535001.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
AK160070.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
BC017637.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
BC025810.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
AK169290.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
AY535001.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
AK160070.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208580 |
BC017637.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109741 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122194 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P137256 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P164703 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P03807730 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P09518447 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P165221 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P09518448 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875908 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875887 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875881 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875889 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875896 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875890 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875899 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875894 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875884 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875880 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875898 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875893 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875868 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875906 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875901 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875867 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875869 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875878 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875902 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875871 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875907 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875886 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875895 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32875872 |
- |
- |