Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027976.2 | NP_082252.1 | 20 - 21 | 2740 | 58 | |
GenBank mRNA | AK006541.1 | - | 9 - 10 | 1217 | 58 | |
AK034942.1 | - | 2740 | 58 | |||
AK145894.1 | - | 20 - 21 | 2401 | 58 | ||
AK150542.1 | - | 20 - 21 | 2406 | 58 | ||
AK150663.1 | - | 20 - 21 | 2406 | 58 | ||
AK151378.1 | - | 20 - 21 | 2195 | 58 | ||
AK161024.1 | - | 20 - 21 | 2093 | 58 | ||
AK168019.1 | - | 8 - 9 | 741 | 58 | ||
BC031544.1 | - | 20 - 21 | 2174 | 3987201 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |