Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tectb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK010948.1 | - | 3 - 4 | 627 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |