Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133216.3 | NP_573479.3 | 8 - 9 | 854 | 66 | |
GenBank mRNA | AF363970.1 | - | 8 - 9 | 794 | 66 | |
AK146033.1 | - | 8 - 9 | 834 | 66 | ||
AK149676.1 | - | 8 - 9 | 793 | 66 | ||
AK158597.1 | - | 6 - 7 | 812 | 66 | ||
AK166715.1 | - | 8 - 9 | 754 | 66 | ||
BC065174.1 | - | 8 - 9 | 742 | 6415314 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |