Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pogz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165948.1 | NP_001159420.1 | 8 - 9 | 1329 | 72 | |
NM_172683.3 | NP_766271.2 | 8 - 9 | 1356 | 72 | ||
GenBank mRNA | AK035255.1 | - | 8 - 9 | 1211 | 72 | |
AK161889.1 | - | 8 - 9 | 1184 | 72 | ||
BC065134.1 | - | 8 - 9 | 1345 | 72 | ||
BC066108.1 | - | 8 - 9 | 1345 | 72 | ||
BC085188.1 | - | 8 - 9 | 1189 | 30682742 | 72 | |
BC096492.1 | - | 7 - 8 | 1199 | 30745658 | 72 | |
BC139432.1 | - | 8 - 9 | 1211 | 8862454 | 72 | |
BC139433.1 | - | 8 - 9 | 1211 | 8862455 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |