Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053198.3 | NP_444428.3 | 3 - 4 | 279 | 70 | |
GenBank mRNA | AF325263.1 | - | 3 - 4 | 221 | 70 | |
AK148530.1 | - | 3 - 4 | 274 | 70 | ||
BC058211.1 | - | 3 - 4 | 276 | 70 | ||
BC089535.1 | - | 3 - 4 | 261 | 6589805 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |