Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053198.3 | NP_444428.3 | 9 - 10 | 552 | 146 | |
GenBank mRNA | AF325263.1 | - | 9 - 10 | 494 | 146 | |
AK148530.1 | - | 8 - 9 | 521 | 146 | ||
BC058211.1 | - | 10 - 11 | 744 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |