Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082484.2 | NP_001075953.1 | 6 - 7 | 1145 | 129 | |
NM_029721.2 | NP_083997.1 | 6 - 7 | 1145 | 129 | ||
GenBank mRNA | AK017836.1 | - | 3 - 4 | 216 | 129 | |
AK122296.1 | - | 6 - 7 | 957 | 129 | ||
AK147452.1 | - | 6 - 7 | 1132 | 129 | ||
BC053495.1 | - | 6 - 7 | 957 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |