Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038139.1 | - | 2 - 3 | 1860 | 73 | |
BC004676.1 | - | 7 - 8 | 975 | 73 | ||
CK392577.1 | - | 2 - 3 | 162 | 73 | ||
X60831.1 | - | 6 - 7 | 709 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |