Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205236.1 | NP_001192165.1 | 6 - 7 | 1912 | 66 | |
NM_133715.5 | NP_598476.2 | 4 - 5 | 881 | 66 | ||
GenBank mRNA | AK046763.1 | - | 4 - 5 | 882 | 66 | |
AK143916.1 | - | 2 - 3 | 353 | 66 | ||
AK154926.1 | - | 6 - 7 | 1913 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |