Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205236.1 | NP_001192165.1 | 7 - 8 | 1997 | 81 | |
NM_133715.5 | NP_598476.2 | 5 - 6 | 966 | 81 | ||
GenBank mRNA | AK046763.1 | - | 5 - 6 | 967 | 81 | |
AK143916.1 | - | 3 - 4 | 438 | 81 | ||
AK154926.1 | - | 7 - 8 | 1998 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |