Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012229.2 | - | 1 - 2 | 661 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |