Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Boll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113367.1 | NP_001106838.1 | 1 - 2 | 228 | 144 | |
GenBank mRNA | AK016403.1 | - | 1 - 2 | 228 | 144 | |
CJ046602.1 | - | 1 - 2 | 228 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |