Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800011776.mm.1 |
NM_001171011.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011776.mm.1 |
NM_001171011.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011776.mm.1 |
TC1800001637.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800011770.mm.1 |
NM_001171010.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011770.mm.1 |
NM_001171010.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011770.mm.1 |
NM_001171011.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011770.mm.1 |
NM_001171011.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011770.mm.1 |
NM_028122.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011770.mm.1 |
NM_028122.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011770.mm.1 |
TC1800001637.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800011769.mm.1 |
NM_001171010.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011769.mm.1 |
NM_001171010.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011769.mm.1 |
NM_001171011.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011769.mm.1 |
NM_001171011.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011769.mm.1 |
NM_028122.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011769.mm.1 |
NM_028122.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011769.mm.1 |
TC1800001637.mm.1 |
MTA TC |