Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122899.1 | NP_001116371.1 | 8 - 9 | 1830 | 102 | |
NM_010704.2 | NP_034834.1 | 8 - 9 | 1830 | 102 | ||
NM_146146.2 | NP_666258.2 | 8 - 9 | 1830 | 102 | ||
GenBank mRNA | AK140785.1 | - | 8 - 9 | 1332 | 102 | |
AK143989.1 | - | 8 - 9 | 1519 | 102 | ||
AK155891.1 | - | 8 - 9 | 1370 | 102 | ||
BC082551.1 | - | 8 - 9 | 1315 | 30621706 | 102 | |
U42467.1 | - | 8 - 9 | 1338 | 102 | ||
U46135.1 | - | 8 - 9 | 1338 | 102 | ||
U49106.1 | - | 7 - 8 | 1278 | 102 | ||
U49109.1 | - | 7 - 8 | 1278 | 102 | ||
U49110.1 | - | 7 - 8 | 1278 | 102 | ||
U52915.1 | - | 8 - 9 | 1792 | 102 | ||
U58861.1 | - | 8 - 9 | 1360 | 102 | ||
U58862.1 | - | 8 - 9 | 1370 | 102 | ||
U58863.1 | - | 8 - 9 | 1360 | 102 | ||
Y10296.1 | - | 7 - 8 | 1286 | 102 | ||
Y10298.1 | - | 7 - 8 | 1286 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |