Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083341.1 | NP_001076810.1 | 10 - 11 | 1189 | 75 | |
NM_026037.3 | NP_080313.2 | 11 - 12 | 1285 | 75 | ||
GenBank mRNA | AB297383.1 | - | 11 - 12 | 1184 | 75 | |
AB305046.1 | - | 11 - 12 | 1184 | 75 | ||
AK003295.1 | - | 2 - 3 | 161 | 75 | ||
AK012931.1 | - | 10 - 11 | 1173 | 75 | ||
AK031824.1 | - | 10 - 11 | 1174 | 75 | ||
AK033106.1 | - | 8 - 9 | 949 | 75 | ||
AK034873.1 | - | 8 - 9 | 973 | 75 | ||
AK076045.1 | - | 10 - 11 | 1179 | 75 | ||
BC025020.1 | - | 10 - 11 | 1139 | 5364068 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |