Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083341.1 | NP_001076810.1 | 8 - 9 | 988 | 99 | |
NM_026037.3 | NP_080313.2 | 9 - 10 | 1084 | 99 | ||
GenBank mRNA | AB297383.1 | - | 9 - 10 | 983 | 99 | |
AB305046.1 | - | 9 - 10 | 983 | 99 | ||
AK012931.1 | - | 8 - 9 | 972 | 99 | ||
AK031824.1 | - | 8 - 9 | 973 | 99 | ||
AK033106.1 | - | 6 - 7 | 748 | 99 | ||
AK034873.1 | - | 6 - 7 | 772 | 99 | ||
AK076045.1 | - | 8 - 9 | 978 | 99 | ||
AK207745.1 | - | 1 - 2 | 82 | 99 | ||
BC025020.1 | - | 8 - 9 | 938 | 5364068 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |