Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145548.4 | NP_663523.2 | 2 - 3 | 447 | 123 | |
GenBank mRNA | AK143903.1 | - | 2 - 3 | 448 | 123 | |
AK143981.1 | - | 2 - 3 | 444 | 123 | ||
AK162358.1 | - | 451 | 123 | |||
AK165405.1 | - | 2 - 3 | 447 | 123 | ||
AK165469.1 | - | 2 - 3 | 425 | 123 | ||
AK165473.1 | - | 2 - 3 | 425 | 123 | ||
BC016446.1 | - | 2 - 3 | 437 | 4235131 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |