Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142950.1 | NP_001136422.1 | 11 - 12 | 1275 | 78 | |
NM_027350.3 | NP_081626.2 | 11 - 12 | 1272 | 78 | ||
GenBank mRNA | AK013880.1 | - | 2 - 3 | 252 | 78 | |
AK077699.1 | - | 11 - 12 | 1253 | 78 | ||
AK157244.1 | - | 11 - 12 | 1235 | 78 | ||
AK172657.1 | - | 11 - 12 | 1234 | 78 | ||
BC005473.1 | - | 11 - 12 | 1178 | 78 | ||
BC052849.1 | - | 11 - 12 | 1233 | 30057822 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |