Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Seh1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039088.1 | NP_001034177.1 | 1 - 2 | 242 | 93 | |
NM_028112.2 | NP_082388.1 | 1 - 2 | 242 | 93 | ||
GenBank mRNA | AK011330.1 | - | 1 - 2 | 144 | 93 | |
AK088835.1 | - | 1 - 2 | 116 | 93 | ||
AK143727.1 | - | 1 - 2 | 242 | 93 | ||
AK169231.1 | - | 1 - 2 | 123 | 93 | ||
AK169388.1 | - | 1 - 2 | 122 | 93 | ||
AK170591.1 | - | 1 - 2 | 195 | 93 | ||
BC027244.1 | - | 1 - 2 | 194 | 3584697 | 93 | |
BY732540.1 | - | 1 - 2 | 242 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |