Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201362.2 | NP_958750.1 | 2 - 3 | 186 | 94 | |
GenBank mRNA | AK078964.1 | - | 2 - 3 | 188 | 94 | |
BC046343.1 | - | 2 - 3 | 166 | 6487853 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |