Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncaip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199151.1 | NP_001186080.1 | 9 - 10 | 1870 | 77 | |
NM_001199153.1 | NP_001186082.1 | 9 - 10 | 1775 | 77 | ||
NM_001199154.1 | NP_001186083.1 | 9 - 10 | 2045 | 77 | ||
NM_026408.4 | NP_080684.3 | 9 - 10 | 1870 | 77 | ||
GenBank mRNA | AF348447.1 | - | 9 - 10 | 1798 | 77 | |
AK013040.1 | - | 7 - 8 | 1604 | 77 | ||
AK017012.1 | - | 6 - 7 | 805 | 77 | ||
AK044974.1 | - | 9 - 10 | 2068 | 77 | ||
AK081942.1 | - | 9 - 10 | 2049 | 77 | ||
AK082079.1 | - | 9 - 10 | 1874 | 77 | ||
AK132346.1 | - | 9 - 10 | 1973 | 77 | ||
BC098489.1 | - | 9 - 10 | 1774 | 30096466 | 77 | |
BC130225.1 | - | 9 - 10 | 1760 | 77 | ||
BC152544.1 | - | 9 - 10 | 1758 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |