Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026386.1 | NP_080662.1 | 8 - 9 | 817 | 72 | |
GenBank mRNA | AK002692.1 | - | 8 - 9 | 817 | 72 | |
AK005470.1 | - | 8 - 9 | 829 | 72 | ||
AK010572.1 | - | 8 - 9 | 814 | 72 | ||
AK075929.1 | - | 8 - 9 | 814 | 72 | ||
AK145840.1 | - | 8 - 9 | 868 | 72 | ||
AK153525.1 | - | 8 - 9 | 814 | 72 | ||
BC006960.1 | - | 8 - 9 | 817 | 2812557 | 72 | |
CT010229.1 | - | 8 - 9 | 802 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |