Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033037.3 | NP_149026.1 | 1 - 2 | 375 | 244 | |
GenBank mRNA | AK004249.1 | - | 1 - 2 | 389 | 244 | |
AK149582.1 | - | 1 - 2 | 389 | 244 | ||
BC013638.1 | - | 1 - 2 | 375 | 4194939 | 244 | |
BC020375.1 | - | 1 - 2 | 375 | 244 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |