Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085373.1 | NP_001078842.1 | 7 - 8 | 1168 | 56 | |
NM_001085374.1 | NP_001078843.1 | 5 - 6 | 1136 | 56 | ||
GenBank mRNA | AK054528.1 | - | 5 - 6 | 1138 | 56 | |
AK158266.1 | - | 7 - 8 | 1169 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |