Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026302.3 | NP_080578.1 | 5 - 6 | 542 | 65 | |
GenBank mRNA | AK016059.1 | - | 5 - 6 | 596 | 65 | |
AK050896.1 | - | 5 - 6 | 546 | 65 | ||
AK160813.1 | - | 5 - 6 | 548 | 65 | ||
AK163240.1 | - | 5 - 6 | 564 | 65 | ||
AK168484.1 | - | 5 - 6 | 548 | 65 | ||
BC006677.1 | - | 5 - 6 | 505 | 65 | ||
BC034725.1 | - | 5 - 6 | 542 | 3667031 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |