Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053129.3 | NP_444359.3 | 1 | 2486 | 82 | |
GenBank mRNA | AK029277.1 | - | 1 | 2478 | 82 | |
AK036169.1 | - | 1 | 2487 | 82 | ||
AK143280.1 | - | 2 | 1813 | 82 | ||
AK160730.1 | - | 1 | 2487 | 82 | ||
AY013786.1 | - | 1 | 2302 | 82 | ||
BC053098.1 | - | 1 | 2459 | 6401988 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |