Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 0610009O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024179.5 | NP_077141.2 | 10 - 11 | 1273 | 95 | |
GenBank mRNA | AK002437.1 | - | 10 - 11 | 1263 | 95 | |
AK009400.1 | - | 3 - 4 | 285 | 95 | ||
AK141675.1 | - | 1236 | 95 | |||
AK171837.1 | - | 10 - 11 | 1264 | 95 | ||
AK172900.1 | - | 10 - 11 | 1198 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |