Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 0610009O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024179.5 | NP_077141.2 | 7 - 8 | 874 | 79 | |
GenBank mRNA | AK002437.1 | - | 7 - 8 | 864 | 79 | |
AK141675.1 | - | 837 | 79 | |||
AK171837.1 | - | 7 - 8 | 865 | 79 | ||
AK172900.1 | - | 7 - 8 | 799 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |