Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198984.1 | NP_001185913.1 | 13 - 14 | 2145 | 110 | |
NM_011552.3 | NP_035682.1 | 13 - 14 | 2145 | 110 | ||
GenBank mRNA | AF001794.1 | - | 13 - 14 | 2089 | 110 | |
AK089518.1 | - | 13 - 14 | 2113 | 110 | ||
BC021514.1 | - | 13 - 14 | 2150 | 5351783 | 110 | |
BC046608.1 | - | 12 - 13 | 1955 | 110 | ||
BC052669.1 | - | 13 - 14 | 2117 | 110 | ||
BC057342.1 | - | 13 - 14 | 2098 | 6825735 | 110 | |
BC060105.1 | - | 13 - 14 | 2098 | 110 | ||
U81030.1 | - | 13 - 14 | 2033 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Transporter Mammalian Gene Collection Mus musculus Probe spans exons |