Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400008570.mm.1 |
NM_026279.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008570.mm.1 |
NM_026279.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008570.mm.1 |
NR_033793.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008570.mm.1 |
NR_033793.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008570.mm.1 |
TC0400001188.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008571.mm.1 |
NM_026279.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008571.mm.1 |
NM_026279.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008571.mm.1 |
NR_033793.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008571.mm.1 |
NR_033793.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008571.mm.1 |
TC0400001188.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008576.mm.1 |
NM_026279.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008576.mm.1 |
NM_026279.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008576.mm.1 |
NR_033793.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008576.mm.1 |
NR_033793.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008576.mm.1 |
TC0400001188.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008573.mm.1 |
NM_026279.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008573.mm.1 |
NM_026279.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008573.mm.1 |
NR_033793.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008573.mm.1 |
NR_033793.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008573.mm.1 |
TC0400001188.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008572.mm.1 |
NM_026279.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008572.mm.1 |
NM_026279.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008572.mm.1 |
NR_033793.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008572.mm.1 |
NR_033793.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008572.mm.1 |
TC0400001188.mm.1 |
MTA TC |