Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037711.2 | NP_001032800.2 | 1 - 2 | 143 | 66 | |
NM_001293727.1 | NP_001280656.1 | 1 - 2 | 143 | 66 | ||
GenBank mRNA | AK018143.1 | - | 1 - 2 | 80 | 66 | |
AK173148.1 | - | 1 - 2 | 120 | 66 | ||
BC042459.1 | - | 1 - 2 | 89 | 66 | ||
BC052941.1 | - | 1 - 2 | 64 | 30021945 | 66 | |
BC064474.1 | - | 1 - 2 | 91 | 66 | ||
BC087546.1 | - | 1 - 2 | 107 | 66 | ||
BC108378.1 | - | 103 | 30736765 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |