Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008945.3 | NP_032971.2 | 1 - 2 | 187 | 64 | |
GenBank mRNA | AK077448.1 | - | 1 - 2 | 164 | 64 | |
AK137044.1 | - | 1 - 2 | 163 | 64 | ||
AK156624.1 | - | 1 - 2 | 163 | 64 | ||
AK161348.1 | - | 1 - 2 | 163 | 64 | ||
AK168340.1 | - | 1 - 2 | 163 | 64 | ||
BC008241.1 | - | 1 - 2 | 142 | 2654421 | 64 | |
CB058553.1 | - | 1 - 2 | 187 | 64 | ||
U65636.1 | - | 1 - 2 | 142 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |