Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081274.1 | NP_001074743.1 | 5 - 6 | 563 | 64 | |
GenBank mRNA | AK002894.1 | - | 5 - 6 | 525 | 64 | |
AK077110.1 | - | 5 - 6 | 525 | 64 | ||
AK145602.1 | - | 5 - 6 | 527 | 64 | ||
AK150210.1 | - | 5 - 6 | 526 | 64 | ||
AK150490.1 | - | 5 - 6 | 522 | 64 | ||
AK151380.1 | - | 5 - 6 | 522 | 64 | ||
AK151993.1 | - | 5 - 6 | 522 | 64 | ||
AK152758.1 | - | 5 - 6 | 522 | 64 | ||
AK152894.1 | - | 5 - 6 | 522 | 64 | ||
AK153409.1 | - | 5 - 6 | 519 | 64 | ||
AK155027.1 | - | 5 - 6 | 522 | 64 | ||
AK161869.1 | - | 5 - 6 | 521 | 64 | ||
AK166733.1 | - | 5 - 6 | 517 | 64 | ||
AK166947.1 | - | 5 - 6 | 499 | 64 | ||
AK167215.1 | - | 5 - 6 | 517 | 64 | ||
AK168251.1 | - | 5 - 6 | 522 | 64 | ||
BC008646.1 | - | 5 - 6 | 494 | 3155147 | 64 | |
BC011329.1 | - | 5 - 6 | 512 | 3984654 | 64 | |
BC014793.1 | - | 5 - 6 | 531 | 3155100 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |