Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172095.1 | NP_001165566.1 | 14 - 15 | 2336 | 80 | |
NM_144787.2 | NP_659036.1 | 14 - 15 | 2400 | 80 | ||
GenBank mRNA | AK144103.1 | - | 14 - 15 | 2363 | 80 | |
AK173023.1 | - | 14 - 15 | 2400 | 80 | ||
BC020180.1 | - | 14 - 15 | 2342 | 4923943 | 80 | |
BC042424.1 | - | 14 - 15 | 2336 | 5357211 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |