Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013783.2 | NP_038811.2 | 14 - 15 | 1689 | 73 | |
GenBank mRNA | AF157105.1 | - | 14 - 15 | 1635 | 73 | |
AF157106.1 | - | 12 - 13 | 1344 | 73 | ||
AF176569.1 | - | 14 - 15 | 1686 | 73 | ||
AF302075.1 | - | 12 - 13 | 1310 | 73 | ||
AF302076.1 | - | 13 - 14 | 1379 | 73 | ||
AF302077.1 | - | 13 - 14 | 1421 | 73 | ||
AK154366.1 | - | 11 - 12 | 1178 | 73 | ||
BC137649.1 | - | 12 - 13 | 1462 | 8860668 | 73 | |
BC144804.1 | - | 12 - 13 | 1462 | 9053352 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |