Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013783.2 | NP_038811.2 | 10 - 11 | 1244 | 64 | |
GenBank mRNA | AF157105.1 | - | 10 - 11 | 1190 | 64 | |
AF157106.1 | - | 8 - 9 | 899 | 64 | ||
AF176569.1 | - | 10 - 11 | 1241 | 64 | ||
AF302075.1 | - | 8 - 9 | 865 | 64 | ||
AF302076.1 | - | 9 - 10 | 934 | 64 | ||
AF302077.1 | - | 8 - 9 | 865 | 64 | ||
AK154366.1 | - | 7 - 8 | 733 | 64 | ||
BC137649.1 | - | 8 - 9 | 1017 | 8860668 | 64 | |
BC144804.1 | - | 8 - 9 | 1017 | 9053352 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |