Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207223.1 | NP_997106.1 | 17 - 18 | 2022 | 66 | |
GenBank mRNA | AK147505.1 | - | 17 - 18 | 2076 | 66 | |
AK159055.1 | - | 14 - 15 | 1655 | 66 | ||
AK185907.1 | - | 3 - 4 | 376 | 66 | ||
AK206031.1 | - | 1 - 2 | 49 | 66 | ||
BC025556.1 | - | 17 - 18 | 1595 | 66 | ||
BC067016.1 | - | 17 - 18 | 2022 | 6827624 | 66 | |
BC098196.1 | - | 17 - 18 | 1598 | 6847993 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |