Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039201.1 | NP_001034290.1 | 3 - 4 | 435 | 92 | |
NM_001039202.1 | NP_001034291.1 | 3 - 4 | 396 | 92 | ||
NM_029826.2 | NP_084102.1 | 3 - 4 | 396 | 92 | ||
GenBank mRNA | AK002837.1 | - | 3 - 4 | 384 | 92 | |
AK014208.1 | - | 3 - 4 | 390 | 92 | ||
AK137285.1 | - | 3 - 4 | 388 | 92 | ||
AK147794.1 | - | 417 | 92 | |||
AK151285.1 | - | 3 - 4 | 384 | 92 | ||
AK152162.1 | - | 3 - 4 | 384 | 92 | ||
AK152259.1 | - | 3 - 4 | 384 | 92 | ||
BC058177.1 | - | 3 - 4 | 370 | 5250739 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |